2-Amino-5-(aminomethyl)naphthalene-1-sulfonic acid 97%

Supplier: Ambeed
Warning

A283796-10G A283796-1G A283796-25G A283796-5G
77262-682EA 31 USD
77262-682 77262-684 77262-686 77262-688 77262-690
2-Amino-5-(aminomethyl)naphthalene-1-sulfonic acid 97%
2-Amino-5-(aminomethyl)naphthalene-1-sulfonic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₃S
MDL Number: MFCD18970027
CAS Number: 52084-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR