3,3'-Oxydianiline 95%

Supplier: Ambeed
Warning

A283606-100MG A283606-1G A283606-250MG A283606-5G
77277-006EA 27 USD
77277-006 77277-008 77277-010 77277-012
3,3'-Oxydianiline 95%
3,3'-Oxydianiline

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O
MDL Number: MFCD09834067
CAS Number: 15268-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR