N1,N3-Dimethyl-N1-(3-(methylamino)propyl)propane-1,3-diamine 98%

Supplier: Ambeed
Danger

A283569-1G A283569-25G A283569-5G
77277-000EA 20 USD
77277-000 77277-002 77277-004
N1,N3-Dimethyl-N1-(3-(methylamino)propyl)propane-1,3-diamine 98%
N1,N3-Dimethyl-N1-(3-(methylamino)propyl)propane-1,3-diamine

Precaución: for Research Use Only

Formula: C₉H₂₃N₃
MW: 173.29 g/mol
MDL Number: MFCD00014857
CAS Number: 123-70-6
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR