1,3-Dimethyl-2-phenyl-2,3-dihydro-1H-benzo[d]imidazole 97%

Supplier: Ambeed
Warning

A283246-100MG A283246-1G A283246-250MG A283246-5G
77262-672EA 14.42 USD
77262-672 77262-674 77262-676 77550-736
1,3-Dimethyl-2-phenyl-2,3-dihydro-1H-benzo[d]imidazole 97%
1,3-Dimethyl-2-phenyl-2,3-dihydro-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₂
MW: 224.30 g/mol
MDL Number: MFCD00186893
CAS Number: 3652-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR