2-Methyl-2-(5-(trifluoromethyl)pyridin-2-yloxy)propanoic acid 95%

Supplier: Ambeed
Warning

A283050-1G A283050-250MG
77262-668EA 327.54 USD
77262-668 77262-670
2-Methyl-2-(5-(trifluoromethyl)pyridin-2-yloxy)propanoic acid 95%
2-Methyl-2-(5-(trifluoromethyl)pyridin-2-yloxy)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₃NO₃
MW: 249.18 g/mol
MDL Number: MFCD11035896
CAS Number: 605680-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR