2'-Amino-3'-hydroxyacetophenone 95%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Amino-3-hydroxyphenyl)ethanone

A282841-1G A282841-25G A282841-5G
77250-530EA 15.45 USD
77250-530 77250-532 77250-534
2'-Amino-3'-hydroxyacetophenone 95%
2'-Amino-3'-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.1626 g/mol
MDL Number: MFCD00191301
CAS Number: 4502-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR