4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic Acid

A282608-100G A282608-10G A282608-1G A282608-25G A282608-500G A282608-5G
77276-988EA 235.87 USD
77276-988 77276-990 77276-992 77276-994 77276-996 77276-998
4-Amino-5-(ethylsulfonyl)-2-methoxybenzoic acid 98%
4-Amino-5-(ethylsulphonyl)-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₅S
MW: 259.28 g/mol
MDL Number: MFCD04973619
CAS Number: 71675-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR