Tributyl(1,3-dioxolan-2-ylmethyl)phosphonium bromide 98%

Supplier: Ambeed
Warning

Synonyms: ((1,3-Dioxolan-2-yl)methyl)tributylphosphonium bromide

A282573-1G A282573-250MG A282573-5G
77250-522EA 14 USD
77250-522 77250-524 77250-526
Tributyl(1,3-dioxolan-2-ylmethyl)phosphonium bromide 98%
Tributyl(1,3-dioxolan-2-ylmethyl)phosphonium bromide

Precaución: for Research Use Only

Formula: C₁₆H₃₄BrO₂P
MDL Number: MFCD09859807
CAS Number: 115754-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR