6-Methylpyridine-2,3-dicarboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A282290-100G
A282290-1G
A282290-250MG
A282290-25G
A282290-5G
77238-898EA
579
USD
77238-898
77238-900
77238-902
77238-904
77238-906
6-Methylpyridine-2,3-dicarboxylic acid 95%
6-Methylpyridine-2,3-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₇NO₄ |
MDL Number:
MFCD00075289 CAS Number: 53636-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...