2,4,6-Trimethylbenzyl alcohol 97%

Supplier: Ambeed
Warning

A282259-100G A282259-1G A282259-250MG A282259-25G A282259-5G
77250-512EA 432 USD
77250-512 77250-514 77250-516 77250-518 77250-520
2,4,6-Trimethylbenzyl alcohol 97%
2,4,6-Trimethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MDL Number: MFCD00014422
CAS Number: 4170-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR