2-(4-Chlorophenoxy)aniline 98%

Supplier: Ambeed
Warning

A281845-10G A281845-1G A281845-5G
77238-886EA 54 USD
77238-886 77238-888 77238-890
2-(4-Chlorophenoxy)aniline 98%
2-(4-Chlorophenoxy)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO
MDL Number: MFCD00025168
CAS Number: 2770-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR