1-Bromo-3-(methoxymethoxy)benzene 98%

Supplier: Ambeed
Warning

A281729-1G A281729-250MG A281729-5G
77238-882EA 36.05 USD
77238-882 77238-884 77550-732
1-Bromo-3-(methoxymethoxy)benzene 98%
1-Bromo-3-(methoxymethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.05 g/mol
MDL Number: MFCD18250001
CAS Number: 42471-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR