Monomethyl phthalate 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Methoxycarbonyl)benzoic acid, Methyl hydrogen phthalate

A281507-100G A281507-10G A281507-25G A281507-5G
77262-658EA 117 USD
77262-658 77262-660 77262-662 77262-664
Monomethyl phthalate 95%
Monomethyl phthalate

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD00002466
CAS Number: 4376-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR