trans-1-Ethoxy-4-(4-pentylcyclohexyl)benzene 97%

Supplier: Ambeed
Warning

A281416-1G A281416-25G A281416-5G
77289-670EA 5 USD
77289-670 77289-672 77289-674
trans-1-Ethoxy-4-(4-pentylcyclohexyl)benzene 97%
trans-1-Ethoxy-4-(4-pentylcyclohexyl)benzene

Precaución: for Research Use Only

Formula: C₁₉H₃₀O
MDL Number: MFCD06658196
CAS Number: 84540-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR