Diethyl 2-vinylcyclopropane-1,1-dicarboxylate 97%

Supplier: Ambeed
Warning

A281359-100MG A281359-1G A281359-250MG
77276-976EA 31.93 USD
77276-976 77276-978 77276-980
Diethyl 2-vinylcyclopropane-1,1-dicarboxylate 97%
Diethyl 2-vinylcyclopropane-1,1-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₄
MW: 212.24 g/mol
MDL Number: MFCD00196937
CAS Number: 7686-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR