2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole 98%

Supplier: Ambeed
Danger

A281276-100MG A281276-250MG
77262-654EA 47.38 USD
77262-654 77262-656
2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole 98%
2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O
MDL Number: MFCD00466332
CAS Number: 33575-83-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR