3-Hydroxy-2-methylquinoline-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A280780-100G A280780-10G A280780-25G
77276-948EA 51.5 USD
77276-948 77276-950 77276-952
3-Hydroxy-2-methylquinoline-4-carboxylic acid 98%
3-Hydroxy-2-methylquinoline-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₃
MW: 203.2 g/mol
MDL Number: MFCD00044850
CAS Number: 117-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR