2,2,2-Trifluoroethyl trifluoromethanesulfonate 97%

Supplier: Ambeed
Danger

A280464-25G A280464-10G A280464-1G A280464-5G
77250-500EA 30 USD
77250-500 77568-566 77593-732 77593-734
2,2,2-Trifluoroethyl trifluoromethanesulfonate 97%
2,2,2-Trifluoroethyl trifluoromethanesulphonate

Precaución: For research use only.

Formula: C₃H₂F₆O₃S
MDL Number: MFCD00671579
CAS Number: 6226-25-1
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR