5-Bromo-2-hydroxy-3-nitropyridine 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-nitro-2-pyridinol, 5-Bromo-3-nitropyridin-2(1H)-one

A280340-100G A280340-10G A280340-25G A280340-5G
77289-654EA 108.15 USD
77289-654 77289-656 77289-658 77748-545
5-Bromo-2-hydroxy-3-nitropyridine 98% (by HPLC)
5-Bromo-2-hydroxy-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₃
MW: 218.99 g/mol
MDL Number: MFCD00023473
CAS Number: 15862-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR