3,3'-Azanediylbis(propan-1-ol) 95%

Supplier: Ambeed
Warning

A280235-100MG A280235-1G A280235-250MG A280235-5G
77276-940EA 17.51 USD
77276-940 77276-942 77276-944 77276-946
3,3'-Azanediylbis(propan-1-ol) 95%
3,3'-Azanediylbis(propan-1-ol)

Precaución: for Research Use Only

Formula: C₆H₁₅NO₂
MW: 133.19 g/mol
MDL Number: MFCD00046013
CAS Number: 14002-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR