1-Hexanesulfonic acid sodium salt 98%

Supplier: Ambeed
Warning

A280199-100G A280199-10G A280199-25G A280199-5G
77238-872EA 58 USD
77238-872 77238-874 77238-876 77238-878
1-Hexanesulfonic acid sodium salt 98%
1-Hexanesulphonic acid sodium salt

Precaución: for Research Use Only

Formula: C₆H₁₃O₃SNa
MDL Number: MFCD00007542
CAS Number: 2832-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR