4-Bromo-1-trityl-1H-Pyrazole 98%

Supplier: Ambeed
Warning

A280030-1G A280030-5G
77289-650EA 16 USD
77289-650 77289-652
4-Bromo-1-trityl-1H-Pyrazole 98%
4-Bromo-1-trityl-1H-Pyrazole

Precaución: for Research Use Only

Formula: C₂₂H₁₇BrN₂
MDL Number: MFCD09907863
CAS Number: 95162-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR