Methyl 1-((4-fluorophenyl)carbamoyl)cyclopropanecarboxylate 97%

Supplier: Ambeed
Warning

A280015-100MG A280015-1G A280015-250MG A280015-5G
77262-644EA 14 USD
77262-644 77262-646 77262-648 77262-650
Methyl 1-((4-fluorophenyl)carbamoyl)cyclopropanecarboxylate 97%
Methyl 1-((4-fluorophenyl)carbamoyl)cyclopropanecarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂FNO₃
MDL Number: MFCD27939981
CAS Number: 1345847-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR