3-Methyl-1H-indole-2-carbonitrile 98%

Supplier: Ambeed
Warning

A279968-100MG A279968-1G A279968-250MG
77238-866EA 73.13 USD
77238-866 77238-868 77238-870
3-Methyl-1H-indole-2-carbonitrile 98%
3-Methyl-1H-indole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈N₂
MW: 156.18 g/mol
MDL Number: MFCD01652234
CAS Number: 13006-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR