5-Amino-1-methyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-pyrazol-5-amine, 5-Amino-1-methylpyrazole

A279893-100G A279893-10G A279893-1G A279893-25G A279893-5G
77289-634EA 138 USD
77289-634 77289-636 77289-638 77289-640 77289-642
5-Amino-1-methyl-1H-pyrazole 98%
5-Amino-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₇N₃
MW: 97.12 g/mol
MDL Number: MFCD00068156
CAS Number: 1192-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR