4-Bromo-5-fluorobenzene-1,2-diamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-5-fluorophenylene-1,2-diamine, 1-Bromo-4,5-diamino-2-fluorobenzene
A279764-100MG
A279764-10G
A279764-1G
A279764-250MG
A279764-25G
A279764-5G
77289-622EA
17.51
USD
77289-622
77289-624
77289-626
77289-628
77289-630
77289-632
4-Bromo-5-fluorobenzene-1,2-diamine 98%
4-Bromo-5-fluorobenzene-1,2-diamine
Precaución: for Research Use Only
Formula:
C₆H₆BrFN₂ MW: 205.028 g/mol |
MDL Number:
MFCD08458150 CAS Number: 153505-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...