2',4'-Dichloropropiophenone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2,4-Dichlorophenyl)-1-propanone, 1-(2,4-Dichlorophenyl)propan-1-one

A279703-100G A279703-10G A279703-25G A279703-5G
77262-636EA 117.42 USD
77262-636 77262-638 77262-640 77262-642
2',4'-Dichloropropiophenone 97%
2',4'-Dichloropropiophenone

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O
MW: 203.07 g/mol
MDL Number: MFCD00027396
CAS Number: 37885-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR