2-(1,3-Benzothiazol-2-yl)acetonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2-(Benzo[d]thiazol-2-yl)acetonitrile

A279680-100G A279680-1G A279680-25G A279680-5G
77262-628EA 229.69 USD
77262-628 77262-630 77262-632 77262-634
2-(1,3-Benzothiazol-2-yl)acetonitrile 95%
2-(1,3-Benzothiazol-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂S
MW: 174.23 g/mol
MDL Number: MFCD00051633
CAS Number: 56278-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR