3',4'-Dimethylacetanilide 97%

Supplier: Ambeed
Warning

Synonyms: N-(3,4-Dimethylphenyl)acetamide

A279664-100G A279664-25G A279664-5G
77276-934EA 147.29 USD
77276-934 77276-936 77276-938
3',4'-Dimethylacetanilide 97%
3',4'-Dimethylacetanilide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO
MW: 163.22 g/mol
MDL Number: MFCD00026137
CAS Number: 2198-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR