4-(Octyloxy)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-n-Octyloxyphenylboronic acid, (4-(Octyloxy)phenyl)boronic acid, 4-n-Octyloxybenzeneboronic acid

A279300-250MG A279300-5G A279300-1G A279300-25G
77289-618EA 5 USD
77289-618 77289-620 77748-540 77748-541
4-(Octyloxy)phenylboronic acid 97%
4-(Octyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₃BO₃
MDL Number: MFCD00982004
CAS Number: 121554-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR