6-Bromo-4-methoxypyrazolo[1,5-a]pyridine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A279183-100MG A279183-1G A279183-250MG
77262-618EA 42 USD
77262-618 77262-620 77616-182
6-Bromo-4-methoxypyrazolo[1,5-a]pyridine-3-carboxylic acid 97%
6-Bromo-4-methoxypyrazolo[1,5-a]pyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₃
MDL Number: MFCD30529017
CAS Number: 2089381-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR