(2,2-Dimethyl-1,3-dioxolan-4-yl)methanamine hydrochloride 96%

Supplier: Ambeed
Warning

A279153-1G A279153-25G A279153-5G
77262-612EA 19.57 USD
77262-612 77262-614 77262-616
(2,2-Dimethyl-1,3-dioxolan-4-yl)methanamine hydrochloride 96%
(2,2-Dimethyl-1,3-dioxolan-4-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂
MW: 167.63 g/mol
MDL Number: MFCD22375314
CAS Number: 167301-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR