(1R,3S,4S)-tert-Butyl 3-(6-bromo-1H-benzo[d]imidazol-2-yl)-2-azabicyclo[2.2.1]heptane-2-carboxylate 95%

Supplier: Ambeed
Warning

A279141-1G A279141-250MG A279141-25G A279141-5G
77238-858EA 30.9 USD
77238-858 77238-860 77238-862 77238-864
(1R,3S,4S)-tert-Butyl 3-(6-bromo-1H-benzo[d]imidazol-2-yl)-2-azabicyclo[2.2.1]heptane-2-carboxylate 95%
(1R,3S,4S)-tert-Butyl 3-(6-bromo-1H-benzo[d]imidazol-2-yl)-2-azabicyclo[2.2.1]heptane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₂BrN₃O₂
MW: 392.29 g/mol
MDL Number: MFCD28386937
CAS Number: 1256387-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR