2-Chloro-N-[2-chloro-5-(trifluoromethyl)phenyl]acetamide 95+%

Supplier: Ambeed
Danger

A278919-100MG A278919-250MG A278919-5G
77262-600EA 8 USD
77262-600 77262-602 77262-604
2-Chloro-N-[2-chloro-5-(trifluoromethyl)phenyl]acetamide 95+%
2-Chloro-N-[2-chloro-5-(trifluoromethyl)phenyl]acetamide

Precaución: for Research Use Only

Formula: C₉H₆Cl₂F₃NO
MDL Number: MFCD00077472
CAS Number: 328-26-7
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR