3-Chloro-5-methoxypyridazine 97%

Supplier: Ambeed
Warning

A278840-1G A278840-250MG A278840-25G A278840-5G
77276-916EA 51.5 USD
77276-916 77276-918 77276-920 77276-922
3-Chloro-5-methoxypyridazine 97%
3-Chloro-5-methoxypyridazine

Precaución: for Research Use Only

Formula: C5H5ClN2O
MW: 144.5600 g/mol
MDL Number: MFCD08236839
CAS Number: 123696-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR