2-Hydrazino-4-(trifluoromethyl)pyrimidine 97%

Supplier: Ambeed
Danger

Synonyms: 2-Hydrazinyl-4-(trifluoromethyl)pyrimidine, [4-(Trifluoromethyl)pyrimidin-2-yl]hydrazine

A278813-1G A278813-25G A278813-5G
77262-588EA 22.66 USD
77262-588 77748-537 77748-538
2-Hydrazino-4-(trifluoromethyl)pyrimidine 97%
2-Hydrazino-4-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₅F₃N₄
MW: 178.118 g/mol
MDL Number: MFCD00115112
CAS Number: 197305-97-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR