(±)-Nipecotic acid 98%

Supplier: Ambeed
Warning

A278584-500G A278584-5G A278584-100G A278584-10G A278584-25G
77289-586EA 164 USD
77289-586 77289-588 77593-716 77616-170 77616-172
(±)-Nipecotic acid 98%
(±)-Nipecotic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MDL Number: MFCD00005992
CAS Number: 498-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR