2-(Methylamino)benzotrifluoride 96%

Supplier: Ambeed
Danger

A278551-1G A278551-250MG A278551-5G
77250-476EA 30 USD
77250-476 77250-478 77250-480
2-(Methylamino)benzotrifluoride 96%
2-(Methylamino)benzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₈F₃N
MDL Number: MFCD00052196
CAS Number: 14925-10-1
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR