3-Bromo-2-chloroquinoline 98%

Supplier: Ambeed
Warning

A278457-10G A278457-1G A278457-250MG
77276-906EA 132 USD
77276-906 77276-908 77276-910
3-Bromo-2-chloroquinoline 98%
3-Bromo-2-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
MDL Number: MFCD08457743
CAS Number: 101870-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR