3-(Chlorocarbonyl)-2-methylphenyl acetate 97%

Supplier: Ambeed
Danger

Synonyms: Acetic Acid 3-(Chlorocarbonyl)-2-methylphenyl Ester, 3-Acetoxy-2-methylbenzoyl Chloride

A278285-25G A278285-5G
77276-902EA 38.11 USD
77276-902 77276-904
3-(Chlorocarbonyl)-2-methylphenyl acetate 97%
3-(Chlorocarbonyl)-2-methylphenyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₃
MW: 212.63 g/mol
MDL Number: MFCD02094037
CAS Number: 167678-46-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR