Benzyl 4-oxoazetidine-2-carboxylate 97%

Supplier: Ambeed
Warning

A277631-100MG A277631-250MG A277631-1G
77289-572EA 24.72 USD
77289-572 77289-574 77593-710
Benzyl 4-oxoazetidine-2-carboxylate 97%
Benzyl 4-oxoazetidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
MDL Number: MFCD00661973
CAS Number: 87791-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR