Benzyl 4-((3-bromophenoxy)methyl)piperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A277465-100MG A277465-250MG
77262-576EA 13.39 USD
77262-576 77262-578
Benzyl 4-((3-bromophenoxy)methyl)piperidine-1-carboxylate 97%
Benzyl 4-((3-bromophenoxy)methyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₂₀H₂₂BrNO₃
MW: 404.29 g/mol
MDL Number: MFCD30609583
CAS Number: 2061979-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR