STF-083010 98%

Supplier: Ambeed
Warning

Synonyms: N-((2-Hydroxynaphthalen-1-yl)methylene)thiophene-2-sulphonamide

A276858-10MG A276858-1MG A276858-50MG A276858-5MG
77238-816EA 69.01 USD
77238-816 77238-818 77238-820 77238-822
STF-083010 98%
STF-083010

Precaución: for Research Use Only

Formula: C₁₅H₁₁NO₃S₂
MW: 317.38 g/mol
MDL Number: MFCD02332975
CAS Number: 307543-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR