Methyl-3-methoxysalicylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-hydroxy-3-methoxybenzoate

A276693-100G A276693-10G A276693-500G A276693-5G A276693-25G
77250-434EA 84 USD
77250-434 77250-436 77250-438 77250-440 77746-493
Methyl-3-methoxysalicylate 97%
Methyl 3-methoxysalicylate

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.1700 g/mol
MDL Number: MFCD00239511
CAS Number: 6342-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR