4-Formyl-N-isopropylbenzamide 95%

Supplier: Ambeed
Warning

A276691-1G A276691-250MG A276691-5G
77289-564EA 15.45 USD
77289-564 77289-566 77748-531
4-Formyl-N-isopropylbenzamide 95%
4-Formyl-N-isopropylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₂
MW: 191.2300 g/mol
MDL Number: MFCD03840703
CAS Number: 13255-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR