Bis(4-iodophenyl)amine 95%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Diiododiphenylamine

A276599-100MG A276599-10G A276599-1G A276599-250MG A276599-5G
77289-550EA 5 USD
77289-550 77289-552 77289-554 77289-556 77289-558
Bis(4-iodophenyl)amine 95%
Bis(4-iodophenyl)amine

Precaución: for Research Use Only

Formula: C₁₂H₉I₂N
MDL Number: MFCD00454093
CAS Number: 20255-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR