4-(4-Methoxyphenyl)-1,3-thiazol-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-(4-methoxyphenyl)-1,3-thiazole, 4-(4-Methoxyphenyl)thiazol-2-amine

A276337-1G A276337-25G
77289-542EA 11.33 USD
77289-542 77289-544
4-(4-Methoxyphenyl)-1,3-thiazol-2-amine 97%
4-(4-Methoxyphenyl)-1,3-thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂OS
MW: 206.27 g/mol
MDL Number: MFCD00170663
CAS Number: 2104-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR