5-(Aminomethyl)-2-methoxyphenol hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-4-methoxybenzylamine hydrochloride, 4-(Aminomethyl)-2-hydroxyanisole hydrochloride

A276221-1G A276221-250MG A276221-5G
77276-866EA 56.65 USD
77276-866 77276-868 77276-870
5-(Aminomethyl)-2-methoxyphenol hydrochloride 97%
5-(Aminomethyl)-2-methoxyphenol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂·ClH
MW: 189.639 g/mol
MDL Number: MFCD05664377
CAS Number: 42365-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR