Methyl-2-hydroxy-2-methoxyacetate 80%

Supplier: Ambeed
Danger

Synonyms: 2-Hydroxy-2-methoxyacetic Acid Methyl Ester, Methyl 2-hydroxy-2-methoxyacetate, Glyoxylic Acid Methyl Ester Methyl Hemiacetal

A276124-25G A276124-5G
77262-566EA 195.7 USD
77262-566 77571-706
Methyl-2-hydroxy-2-methoxyacetate 80%
Methyl-2-hydroxy-2-methoxyacetate

Precaución: for Research Use Only

Formula: C₄H₈O₄
MW: 120.10 g/mol
MDL Number: MFCD00043941
CAS Number: 19757-97-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR