Pibrentasvir ≥98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl ((2S,2'S,3R,3'R)-((2S,2'S)-(((2R,5R)-1-(3,5-difluoro-4-(4-(4-fluorophenyl)piperidin-1-yl)phenyl)pyrrolidine-2,5-diyl)bis(5-fluoro-1H-benzo[d]imidazole-6,2-diyl))bis(pyrrolidine-2,1-diyl))bis(3-methoxy-1-oxobutane-1,2-diyl))dicarbamate, ABT-530, Dimethyl ((2S,2'S,3R,3'R)-((2S,2'S)-(((2R,5R)-1-(3,5-difluoro-4-(4-(4-fluorophenyl)piperidin-1-yl)phenyl)pyrrolidine-2,5-diyl)bis(6-fluoro-1H-benzo[d]imidazole-5,2-diyl))bis(pyrrolidine-2,1-diyl))bis(3-methoxy-1-oxobutane-1,2-diyl))dicarbamate

A275632-10MG A275632-1MG A275632-25MG A275632-5MG
77262-550EA 46.35 USD
77262-550 77262-552 77262-554 77262-556
Pibrentasvir ≥98%
Pibrentasvir

Precaución: for Research Use Only

Formula: C₅₇H₆₅F₅N₁₀O₈
MW: 1113.2 g/mol
MDL Number: MFCD30747902
CAS Number: 1353900-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR